siRNA Details
Gene Symbol: Dock6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108997.1 | NP_001102467.1 | 42 | 5380 | ||
XM_006242657.2 | XP_006242719.1 | 44 | 5515 | |||
XM_006242658.2 | XP_006242720.1 | 43 | 5422 | |||
XM_006242659.2 | XP_006242721.1 | 43 | 5410 | |||
XM_008765964.1 | XP_008764186.1 | 43 | 5452 | |||
XM_008765965.1 | XP_008764187.1 | 43 | 5440 | |||
XM_008765966.1 | XP_008764188.1 | 42 | 5401 |
