siRNA Details
Gene Symbol: Arhgef12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013246.1 | NP_001013264.1 | 15 | 1269 | ||
GenBank mRNA | AY935257.1 | - | 15 | 1269 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arhgef12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013246.1 | NP_001013264.1 | 15 | 1269 | ||
GenBank mRNA | AY935257.1 | - | 15 | 1269 |