siRNA Details
Gene Symbol: Arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109001.1 | NP_001102471.1 | 4 | 790 | ||
XM_006243171.2 | XP_006243233.1 | 5 | 887 | |||
XM_008766323.1 | XP_008764545.1 | 5 | 3034 |
