siRNA Details
Gene Symbol: Cpne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109003.1 | NP_001102473.1 | 14 | 1812 | ||
XM_008766557.1 | XP_008764779.1 | 11 | 1427 | |||
GenBank mRNA | AM747281.1 | - | 10 | 975 |
