siRNA Details
Gene Symbol: Fam135a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134597.1 | NP_001128069.1 | 15 | 2743 | ||
XM_006244664.1 | XP_006244726.1 | 14 | 2780 | |||
XM_006244665.1 | XP_006244727.1 | 16 | 2451 | |||
XM_006244666.1 | XP_006244728.1 | 13 | 2114 | |||
GenBank mRNA | FQ222914.1 | - | 16 | 2451 |
