siRNA Details
Gene Symbol: Stat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012226.1 | NP_001012226.1 | 4, 5 | 852 | ||
GenBank mRNA | BC081938.1 | - | 4, 5 | 852 | 7115387 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012226.1 | NP_001012226.1 | 4, 5 | 852 | ||
GenBank mRNA | BC081938.1 | - | 4, 5 | 852 | 7115387 |