siRNA Details
Gene Symbol: Ldhal6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183334.1 | NP_899163.1 | 1 | 267 | ||
GenBank mRNA | AY342428.1 | - | 1 | 267 | ||
BC078970.1 | - | 1 | 248 | 7111795 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ldhal6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183334.1 | NP_899163.1 | 1 | 267 | ||
GenBank mRNA | AY342428.1 | - | 1 | 267 | ||
BC078970.1 | - | 1 | 248 | 7111795 |