siRNA Details
Gene Symbol: Olr786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000916.1 | NP_001000916.1 | 1 | 731 |
Gene Symbol: LOC100909613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749539.1 | XP_003749587.1 | 731 |
