siRNA Details
Gene Symbol: Wnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002823.2 | NP_001002823.2 | 9 | 2351 | ||
NM_001199095.1 | NP_001186024.1 | 10 | 2609 | |||
XM_008763215.1 | XP_008761437.1 | 9 | 2417 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001199083, NM_001199084, BC146009, BC146011.
Gene Ontology Categories:
