siRNA Details
Gene Symbol: C5ar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003710.2 | NP_001003710.2 | 2 | 758 | ||
XM_006228352.2 | XP_006228414.1 | 2 | 1140 | |||
XM_006228353.2 | XP_006228415.1 | 2 | 912 | |||
GenBank mRNA | AY600435.1 | - | 1 | 624 | ||
AY741660.1 | - | 2 | 758 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
