siRNA Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271089.2 | NP_001258018.1 | 5 | 881 | ||
NM_019283.3 | NP_062156.2 | 6 | 1006 | |||
GenBank mRNA | AB015433.1 | - | 6 | 972 | ||
BC061989.1 | - | 5 | 864 | 5599495 | ||
CV109741.1 | - | 4 | 223 | |||
U59324.1 | - | 5 | 795 |
