siRNA Details
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017315.2 | NP_059011.1 | 14 | 1730 | ||
XM_006254602.2 | XP_006254664.1 | 16 | 2111 | |||
GenBank mRNA | AF080452.1 | - | 14 | 1730 | ||
BC078851.1 | - | 14 | 1730 | 7127602 |
