siRNA Details
Gene Symbol: Ptk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017318.2 | NP_059014.2 | 17 | 1589 | ||
XM_006252143.2 | XP_006252205.1 | 17 | 1914 | |||
XM_006252144.2 | XP_006252206.1 | 17 | 4905 | |||
XM_006252145.2 | XP_006252207.1 | 17 | 1718 | |||
XM_008770773.1 | XP_008768995.1 | 17 | 3102 | |||
XM_008770774.1 | XP_008768996.1 | 17 | 2668 | |||
GenBank mRNA | BC101921.1 | - | 17 | 1589 | 7937929 | |
D45854.1 | - | 17 | 1767 | |||
U69109.1 | - | 16 | 1509 |
