siRNA Details
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019309.2 | NP_062182.1 | 7 | 1357 | ||
GenBank mRNA | Z11548.1 | - | 1377 | |||
Z11715.1 | - | 7 | 1155 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019309.2 | NP_062182.1 | 7 | 1357 | ||
GenBank mRNA | Z11548.1 | - | 1377 | |||
Z11715.1 | - | 7 | 1155 |