siRNA Details
Gene Symbol: Rgs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019339.1 | NP_062212.1 | 1 | 846 | ||
XM_006251351.2 | XP_006251413.1 | 2 | 978 | |||
XM_006251352.2 | XP_006251414.1 | 2 | 982 | |||
XM_006251353.2 | XP_006251415.1 | 2 | 1502 | |||
XM_008770350.1 | XP_008768572.1 | 2 | 1515 | |||
GenBank mRNA | U92280.1 | - | 1 | 846 |
