siRNA Details
Gene Symbol: Slc14a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019346.2 | NP_062219.2 | 6, 7 | 929 | ||
XM_008772168.1 | XP_008770390.1 | 7, 8 | 1147 | |||
XM_008772169.1 | XP_008770391.1 | 6, 7 | 2061 | |||
XM_008772170.1 | XP_008770392.1 | 6, 7 | 998 | |||
GenBank mRNA | FQ211495.1 | - | 6, 7 | 962 | ||
X98399.1 | - | 6, 7 | 929 |
