siRNA Details
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019359.1 | NP_062232.1 | 7 | 1176 | ||
GenBank mRNA | BC062020.1 | - | 7 | 1179 | 5623281 | |
FQ209647.1 | - | 2 | 399 | |||
FQ228936.1 | - | 4 | 613 | |||
U06755.1 | - | 7 | 1176 |
