siRNA Details
Gene Symbol: Itih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019369.2 | NP_062242.2 | 10 | 1271 | ||
GenBank mRNA | BC089806.1 | - | 10 | 1271 | 7326076 | |
FQ210475.1 | - | 10 | 1227 | |||
Y11283.1 | - | 10 | 1268 |
