siRNA Details
Gene Symbol: Enpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019370.2 | NP_062243.2 | 10 | 945 | ||
GenBank mRNA | AJ250374.1 | - | 6 | 439 | ||
BC097326.1 | - | 10 | 945 | 7458431 | ||
D30649.1 | - | 10 | 972 | |||
U78787.1 | - | 10 | 942 | |||
U78788.1 | - | 10 | 948 | |||
Z47987.1 | - | 10 | 929 |
Gene Symbol: LOC103690935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758654.1 | XP_008756876.1 | 5 | 728 |
