siRNA Details
Gene Symbol: Pdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271108.2 | NP_001258037.1 | 3 | 619 | ||
NM_019372.4 | NP_062245.2 | 2 | 614 | |||
XM_006237907.2 | XP_006237969.1 | 2 | 734 | |||
XM_006237908.2 | XP_006237970.1 | 2 | 607 | |||
XM_006237909.2 | XP_006237971.1 | 3 | 698 | |||
GenBank mRNA | AF062740.1 | - | 2 | 596 | ||
BC129095.1 | - | 2 | 605 | 8369880 | ||
FN800686.1 | - | 2 | 614 |
