siRNA Details
Gene Symbol: Pdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271108.2 | NP_001258037.1 | 3 | 665 | ||
NM_019372.4 | NP_062245.2 | 2 | 660 | |||
XM_006237907.2 | XP_006237969.1 | 2 | 780 | |||
XM_006237908.2 | XP_006237970.1 | 2 | 653 | |||
XM_006237909.2 | XP_006237971.1 | 3 | 744 | |||
GenBank mRNA | AF062740.1 | - | 2 | 642 | ||
BC129095.1 | - | 2 | 651 | 8369880 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001098230, NM_001098231, NM_001033453, BC048372, AK133542, and others.
Gene Ontology Categories:
