siRNA Details
Gene Symbol: Hgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019387.2 | NP_062260.2 | 16 | 1428 | ||
XM_006247908.2 | XP_006247970.1 | 16 | 1609 | |||
XM_006247909.2 | XP_006247971.1 | 16 | 1606 | |||
GenBank mRNA | AB002811.1 | - | 16 | 1434 | ||
AF036344.1 | - | 16 | 1464 | |||
BC083561.1 | - | 16 | 1428 | 7133877 | ||
U87863.1 | - | 16 | 1411 |
