siRNA Details
Gene Symbol: Ifit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020096.1 | NP_064481.1 | 1 | 1255 | ||
GenBank mRNA | AJ276893.1 | - | 1 | 1255 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020096.1 | NP_064481.1 | 1 | 1255 | ||
GenBank mRNA | AJ276893.1 | - | 1 | 1255 |