siRNA Details
Gene Symbol: Adap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020101.1 | NP_064486.1 | 8 | 820 | ||
XM_008768109.1 | XP_008766331.1 | 9 | 1259 | |||
GenBank mRNA | AJ238993.1 | - | 8 | 820 | ||
FQ225344.1 | - | 2 | 70 |
