siRNA Details
Gene Symbol: Adam18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253341.2 | XP_006253403.1 | 11 | 1009 | ||
XM_008771369.1 | XP_008769591.1 | 11 | 1009 | |||
GenBank mRNA | Y11490.1 | - | 2 | 118 |
