siRNA Details
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021265.3 | NP_067088.2 | 9 | 832 | ||
XM_006245011.2 | XP_006245073.1 | 10 | 2119 | |||
XM_008767130.1 | XP_008765352.1 | 10 | 2119 | |||
XM_008767131.1 | XP_008765353.1 | 11 | 1259 | |||
XM_008767132.1 | XP_008765354.1 | 12 | 1145 | |||
XM_008767133.1 | XP_008765355.1 | 8 | 714 | |||
GenBank mRNA | AB030499.1 | - | 7 | 781 | ||
AB030500.1 | - | 7 | 781 | |||
AB030501.1 | - | 7 | 781 | |||
BF399374.1 | - | 7 | 413 |
