siRNA Details
Gene Symbol: Nup210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053322.2 | NP_445774.1 | 5 | 576 | ||
XM_006236921.2 | XP_006236983.1 | 3 | 757 | |||
GenBank mRNA | Y00826.1 | - | 5 | 577 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nup210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053322.2 | NP_445774.1 | 5 | 576 | ||
XM_006236921.2 | XP_006236983.1 | 3 | 757 | |||
GenBank mRNA | Y00826.1 | - | 5 | 577 |