siRNA Details
Gene Symbol: Ltbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021586.2 | NP_067597.2 | 7 | 1498 | ||
GenBank mRNA | Y12760.1 | - | 7 | 1497 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ltbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021586.2 | NP_067597.2 | 7 | 1498 | ||
GenBank mRNA | Y12760.1 | - | 7 | 1497 |