siRNA Details
Gene Symbol: Ambra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134341.1 | NP_001127813.1 | 7 | 2364 | ||
XM_008762029.1 | XP_008760251.1 | 7 | 2551 | |||
XM_008762030.1 | XP_008760252.1 | 7 | 2268 | |||
XM_008762031.1 | XP_008760253.1 | 6 | 2125 | |||
XM_008762032.1 | XP_008760254.1 | 6 | 2125 | |||
XM_008762033.1 | XP_008760255.1 | 7 | 1852 | |||
XM_008762034.1 | XP_008760256.1 | 7 | 1852 | |||
GenBank mRNA | AB027149.1 | - | 1 | 439 |
