siRNA Details
Gene Symbol: Erbb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021687.1 | NP_067719.1 | 28 | 3567 | ||
GenBank mRNA | AF041838.1 | - | 28 | 3567 | ||
AY375306.1 | - | 28 | 3534 | |||
AY375307.1 | - | 27 | 3486 | |||
U52531.1 | - | 4 | 444 |
