siRNA Details
Gene Symbol: Trpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021771.2 | NP_068539.2 | 9 | 2399 | ||
XM_006232282.2 | XP_006232344.2 | 9 | 2564 | |||
XM_008760941.1 | XP_008759163.1 | 9 | 2564 | |||
GenBank mRNA | JN160741.1 | - | 9 | 2160 |
