siRNA Details
Gene Symbol: Rhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022505.2 | NP_071950.1 | 7 | 1078 | ||
XM_006239268.2 | XP_006239330.1 | 6 | 974 | |||
GenBank mRNA | AB015191.1 | - | 7 | 1043 | ||
AF531096.1 | - | 7 | 1078 | |||
BC127471.1 | - | 7 | 1067 | 8374048 |
