siRNA Details
Gene Symbol: Tsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021854.1 | NP_068626.1 | 4 | 460 | ||
XM_006233843.2 | XP_006233905.1 | 6 | 914 | |||
XM_006233844.2 | XP_006233906.1 | 6 | 885 | |||
XM_006233845.2 | XP_006233907.1 | 6 | 736 | |||
XM_006233846.2 | XP_006233908.1 | 6 | 3001 | |||
XM_006233847.2 | XP_006233909.1 | 6 | 1222 | |||
XM_006233849.2 | XP_006233911.1 | 6 | 914 | |||
GenBank mRNA | AB011821.1 | - | 4 | 460 |
