siRNA Details
Gene Symbol: Clock | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021856.2 | NP_068628.1 | 15 | 1469 | ||
XM_006250869.2 | XP_006250931.1 | 15 | 1513 | |||
XM_006250870.2 | XP_006250932.1 | 15 | 1513 | |||
GenBank mRNA | AB019258.1 | - | 15 | 1469 |
