siRNA Details
Gene Symbol: Clock | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289832.1 | NP_001276761.1 | 16 | 1777 | ||
NM_021856.2 | NP_068628.1 | 17 | 1867 | |||
XM_006250869.2 | XP_006250931.1 | 17 | 1911 | |||
XM_006250870.2 | XP_006250932.1 | 17 | 1911 | |||
GenBank mRNA | AB019258.1 | - | 17 | 1867 | ||
AB019259.1 | - | 16 | 1777 |
