siRNA Details
Gene Symbol: Hspa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021863.3 | NP_068635.1 | 1 | 1706 | ||
XM_006240247.2 | XP_006240309.1 | 2 | 1683 | |||
GenBank mRNA | BC081803.1 | - | 2 | 1677 | 7112083 |
