siRNA Details
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022282.1 | NP_071618.1 | 14, 15 | 1899 | ||
XM_008759648.1 | XP_008757870.1 | 14, 15 | 2046 | |||
XM_008759649.1 | XP_008757871.1 | 2, 3 | 2484 | |||
GenBank mRNA | U49049.1 | - | 14, 15 | 1899 | ||
U50717.1 | - | 14, 15 | 1605 |
