siRNA Details
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022282.1 | NP_071618.1 | 5, 6 | 710 | ||
XM_008759648.1 | XP_008757870.1 | 5, 6 | 857 | |||
GenBank mRNA | U49049.1 | - | 5, 6 | 710 | ||
U50717.1 | - | 5, 6 | 434 |
