siRNA Details
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022289.3 | NP_071625.2 | 7 | 1125 | ||
GenBank mRNA | AF068746.2 | - | 6 | 837 | ||
AF305780.1 | - | 7 | 1111 | |||
BC061554.1 | - | 7 | 1129 | 6890629 |
