siRNA Details
Gene Symbol: Pi4ka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022301.1 | NP_071637.1 | 25, 26 | 2907 | ||
XM_008768902.1 | XP_008767124.1 | 25, 26 | 3060 | |||
GenBank mRNA | D83538.1 | - | 25, 26 | 3185 |
