siRNA Details
Gene Symbol: Mafg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022386.2 | NP_071781.1 | 3 | 639 | ||
XM_006247912.2 | XP_006247974.1 | 3 | 4722 | |||
XM_006247913.2 | XP_006247975.1 | 4 | 3723 | |||
XM_006247914.2 | XP_006247976.1 | 2 | 642 | |||
XM_006247916.2 | XP_006247978.1 | 2 | 2419 | |||
XM_006247917.2 | XP_006247979.1 | 2 | 5191 | |||
XM_006247918.2 | XP_006247980.1 | 2 | 1802 | |||
XM_006247919.2 | XP_006247981.1 | 2 | 2349 | |||
XM_006247921.2 | XP_006247983.1 | 3 | 1400 | |||
XM_006247922.2 | XP_006247984.1 | 3 | 561 | |||
XM_006247923.2 | XP_006247985.1 | 3 | 1330 | |||
XM_008768482.1 | XP_008766704.1 | 3 | 4172 | |||
GenBank mRNA | AB026487.1 | - | 2 | 513 | ||
AB050011.1 | - | 1 | 566 | |||
BC078828.1 | - | 3 | 639 | 7127046 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010756, BC052633, AB221541, BC002092.
Gene Ontology Categories:
