siRNA Details
Gene Symbol: Mafg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022386.2 | NP_071781.1 | 3 | 288 | ||
XM_006247912.2 | XP_006247974.1 | 3 | 4371 | |||
XM_006247913.2 | XP_006247975.1 | 4 | 3372 | |||
XM_006247914.2 | XP_006247976.1 | 2 | 291 | |||
XM_006247916.2 | XP_006247978.1 | 2 | 2068 | |||
XM_006247917.2 | XP_006247979.1 | 2 | 4840 | |||
XM_006247918.2 | XP_006247980.1 | 2 | 1451 | |||
XM_006247919.2 | XP_006247981.1 | 2 | 1998 | |||
XM_006247921.2 | XP_006247983.1 | 3 | 1049 | |||
XM_006247922.2 | XP_006247984.1 | 3 | 210 | |||
XM_006247923.2 | XP_006247985.1 | 3 | 979 | |||
XM_008768482.1 | XP_008766704.1 | 3 | 3821 | |||
GenBank mRNA | AB026487.1 | - | 2 | 162 | ||
AB050011.1 | - | 1 | 215 | |||
BC078828.1 | - | 3 | 288 | 7127046 |
