siRNA Details
Gene Symbol: Pdlim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053326.1 | NP_445778.1 | 5 | 600 | ||
XM_006233404.2 | XP_006233466.1 | 5 | 1341 | |||
XM_006233405.2 | XP_006233467.1 | 5 | 1339 | |||
XM_006233406.2 | XP_006233468.1 | 5 | 1337 | |||
XM_006233407.2 | XP_006233469.1 | 5 | 1336 | |||
XM_006233412.2 | XP_006233474.1 | 5 | 1328 | |||
XM_006233416.2 | XP_006233478.1 | 5 | 1327 | |||
XM_006233420.2 | XP_006233482.1 | 5 | 1059 | |||
GenBank mRNA | U48247.1 | - | 5 | 600 |
