siRNA Details
Gene Symbol: Pdlim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053326.1 | NP_445778.1 | 8, 9 | 1158 | ||
XM_006233407.2 | XP_006233469.1 | 13, 14 | 2323 | |||
XM_006233414.2 | XP_006233476.1 | 12, 13 | 1975 | |||
XM_006233415.2 | XP_006233477.1 | 11, 12 | 1956 | |||
XM_006233418.2 | XP_006233480.1 | 9, 10 | 1392 | |||
XM_006233419.2 | XP_006233481.1 | 8, 9 | 1372 | |||
GenBank mRNA | U48247.1 | - | 8, 9 | 1158 |
