siRNA Details
Gene Symbol: Ech1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022594.1 | NP_072116.1 | 4 | 444 | ||
GenBank mRNA | BC060565.1 | - | 4 | 450 | ||
BC062226.1 | - | 4 | 450 | 6890529 | ||
FQ214121.1 | - | 4 | 446 | |||
FQ230435.1 | - | 4 | 457 | |||
U08976.1 | - | 4 | 444 |
