siRNA Details
Gene Symbol: Ilkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022606.1 | NP_072128.1 | 11 | 1025 | ||
XM_008767296.1 | XP_008765518.1 | 10 | 2981 | |||
XM_008767297.1 | XP_008765519.1 | 9 | 3306 | |||
GenBank mRNA | AF095927.1 | - | 11 | 1025 | ||
BC062010.1 | - | 11 | 1027 | 5621497 |
