siRNA Details
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022608.1 | NP_072130.1 | 9 | 706 | ||
GenBank mRNA | AJ132352.1 | - | 9 | 721 | ||
BC162038.1 | - | 9 | 706 | 7099625 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022608.1 | NP_072130.1 | 9 | 706 | ||
GenBank mRNA | AJ132352.1 | - | 9 | 721 | ||
BC162038.1 | - | 9 | 706 | 7099625 |