siRNA Details
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113740.1 | NP_001107212.1 | 8, 9 | 843 | ||
NM_022616.2 | NP_072138.2 | 7, 8 | 840 | |||
GenBank mRNA | AF142759.1 | - | 7, 8 | 807 | ||
BC128738.1 | - | 8, 9 | 741 | 7381331 |
Gene Symbol: LOC100910754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772553.1 | XP_008770775.1 | 1 | 832 |
Gene Symbol: LOC102550225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772341.1 | XP_008770563.1 | 5 | 874 |
