siRNA Details
Gene Symbol: Exoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022691.1 | NP_073182.1 | 12, 13 | 1701 | ||
GenBank mRNA | AF032667.1 | - | 12, 13 | 1701 | ||
FQ231273.1 | - | 1468 |
Gene Symbol: LOC100911734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768426.1 | XP_008766648.1 | 14, 15 | 1614 | ||
XM_008768427.1 | XP_008766649.1 | 13, 14 | 1575 | |||
XM_008768428.1 | XP_008766650.1 | 13, 14 | 1521 | |||
XM_008768429.1 | XP_008766651.1 | 12, 13 | 1482 | |||
XM_008768430.1 | XP_008766652.1 | 1743 |
