siRNA Details
Gene Symbol: Taok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022702.1 | NP_073193.1 | 16 | 2608 | ||
XM_006230329.2 | XP_006230391.1 | 16 | 4049 | |||
XM_006230330.2 | XP_006230392.1 | 15 | 2470 | |||
GenBank mRNA | AF140556.2 | - | 16 | 2608 |
